ContactUsLinkComponent
HIS Lite™ iFluor® 568 Tris NTA Chelator
HIS Lite™ iFluor® 568 Tris NTA Chelator
N° de catalogue AATB12657
Fournisseur:  AAT BIOQUEST
New Product
Numéro CAS:  
HIS Lite™ iFluor® 568 Tris NTA Chelator
N° de catalogue AATB12657
Fournisseur:  AAT BIOQUEST
New Product
Numéro CAS:  
Spécifications
  • Cdt:
    100 µG
  • Désignation:
    HIS Lite™ iFluor™ 568 tris NTA chelator
Spécifications
À propos de cet article
iFluor® 568 Tris-NTA Chelator is used as a sensitive fluorescent probe for detecting polyhistidine-labelled proteins in combination with the addition of a heavy metal ion (such as Ni²⁺) in cells, solution and solid surfaces.
  • Stable for 24 months
Quality tested in-house.
Attention: For research use only.