Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
XFD488 alkyne
XFD488 alkyne, FastClick™
XFD488 alkyne, FastClick™
XFD488 alkyne, FastClick™
Catalog # AATB72875
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
XFD488 alkyne, FastClick™
Catalog # AATB72875
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Cdt:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
773.92 g/mol
Product Details