Anti-LEF1 Mouse Recombinant Antibody [clone: rLEF1/6906]
ANTIA277907-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:DKFZp586H0919
- Symbole de l'antigène:LEF1
- Clonalité:Recombinant
- Clone:rLEF1/6906
- Conjugaison:Unconjugated
- Hôte:Mouse
- ImmunoChimie:Yes
- Isotype:IgG1 kappa
- Réactivité:Human
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# Q9UJU2
- Synonymes antigène:T cell specific transcription factor 1 alpha|FLJ46390|LEF1_HUMAN|LEF-1|T cell-specific transcription factor 1-alpha|LEF1|Lymphoid enhancer binding factor 1|LEF 1|Lymphoid enhancer-binding factor 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:44 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Recombinant fragment, around amino acids 100-200, of human LEF1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Recombinant mouse monoclonal [rLEF1/6906] antibody to LEF1 for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: LEF1
Clonality: Recombinant
Clone: rLEF1/6906
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human