ReadiCleave™ AML Cleavage Buffer, 5X Aqueous Solution, AAT Bioquest
ReadiCleave™ AML cleavage buffer 5X aqueous solution, 1 mL
Catalog # AATB7510
Supplier: AAT Bioquest
ReadiCleave™ AML Cleavage Buffer, 5X Aqueous Solution, AAT Bioquest
Catalog # AATB7510
Supplier: AAT Bioquest
CAS Number:
Jump to:
Product Details & Documents
ReadiCleave™ AML Cleavage Buffer provides an optimised formula to effectively remove the fluorescence tags from ReadiCleave™ AML-labeled biological targets when needed.
- Designed for ease of use
- Saves time
The removal of the fluorescent ReadiCleave™ AML tags may make the fluorescence imaging more compatible with the downstream analysis of a biological target by other techniques, e.g., MS analysis.
Caution: For research use only.
Specifications
Specifications
- Cdt:1 mL
- Désignation:ReadiCleave™ AML cleavage buffer 5X aqueous solution