ReadiUse™ PCR Reaction Buffer
ReadiUse™ PCR Reaction Buffer
ReadiUse™ PCR reaction buffer, 50 mL
N° de catalogue AATB17202
Fournisseur:   AAT Bioquest
ReadiUse™ PCR Reaction Buffer
N° de catalogue AATB17202
Fournisseur:  AAT Bioquest
Numéro CAS:  
ReadiUse™ PCR Reaction Buffer
ReadiUse™ PCR Reaction Buffer
ReadiUse™ PCR reaction buffer, 50 mL
Numéro CAS:  
Prix
135.00 SFr.
Connexion Pour la disponibilité
1 * 50 mL
Product Details & Documents
This ready to use PCR buffer contains several components essential for the PCR reaction.
  • Designed for ease of use
  • MgCl2 (magnesium chloride), a cofactor, is used for maintaining the activity of the DNA polymerase enzyme
PCR (polymerase chain reaction) is a laboratory technique used to amplify DNA (deoxyribonucleic acid) segments.
Caution: For research use only.
Spécifications
Spécifications
  • Cdt:
    50 mL
  • Désignation:
    ReadiUse™ PCR reaction buffer