CMV (pp65) Peptide Pool
N° de catalogue STMC100-0668
Fournisseur: STEMCELL Technologies
Numéro CAS:
Spécifications
- Cdt:1 KIT
- Seq:0075
- Nom de la protéine/du peptide:CMV (pp65) Peptide Pool
- Purity:Average 70%
Spécifications
À propos de cet article
The CMV (pp65) peptide pool is a lyophilised mixture of 138 peptides from the 65 kDa phosphoprotein (pp65) of human cytomegalovirus (CMV; strain AD169). pp65 has been shown to counteract the induction of interferon response by preventing the activation of interferon response factor 3 (IRF-3). pp65 is also important in inducing transcription of viral immediate-early promoter. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 561 on pp65.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').