ContactUsLinkComponent
Dual Position Cap for Culture Tubes
  LBCN3500-800-000-9
 :  LABCON
New Product
Dual Position Cap for Culture Tubes
  LBCN3500-800-000-9
 :  LABCON
New Product
  • Emballage:
    Packed in bags
  • Désignation:
    Dual position caps for 17×100 mm culture tubes
  • Cdt:
    1.000
Tube caps are made of low density polyethylene for a secure seal and are available in a fluted plug style or dual position design that permits sample aspiration.
Polyethylene caps are not autoclavable, but are available sterile with many of the tube packages.