Silane-Prep Slides
Silane prep slides, Verre
Catalog # S4651-72EA
Supplier: SIGMA ALDRICH MICROSCOPY
Jump to:
Product Details & Documents
Silane-prep slides are microscope slides coated with silane (aminoalkysilane) to improve the adhesion of tissues to the slide for immunohistochemistry. The slide includes a 'frosted end' for slide/patient identification. It is not tested for use in microarray assays.
- Silane-prep slides have been used in
- Immunofluorescence microscopy of DNA fibers
- Immunohistochemistry
- The determination of hybridisation stringency
- Hematology and histology
- Biological source: synthetic
Specifications
- Désignation:Silane prep slides
- Matière:Verre
- Cat. No.:S4651-72EA