ContactUsLinkComponent
STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit
STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit
Référence STMC05010
Fournisseur:  STEMCELL Technologies
Numéro CAS:  
undefined
STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit
Référence STMC05010
Fournisseur:  STEMCELL Technologies
Numéro CAS:  
Spécifications
  • Cdt:
    1 KIT
  • Désignation:
    STEMdiff™ ventricular cardiomyocyte differentiation kit
Spécifications
À propos de cet article
Serum-free media for differentiation of human PSCs to cardiomyocytes and long-term maintenance of human PSC-derived cardiomyocytes.
STEMdiff™ ventricular cardiomyocyte differentiation kit includes a medium for differentiation of human embryonic stem (ES) and induced pluripotent stem (iPS) cells (human pluripotent stem cells [hPSCs]) into ventricular cardiomyocytes (cardiac troponin T-positive [cTnT+]), as well as a medium for maintenance of hPSC-derived cardiomyocytes. This serum-free kit can be used to generate ventricular cardiomyocytes derived from a clump culture of hPSCs maintained in mTeSR™1, mTeSR™ plus, TeSR™-AOF, or TeSR™-E8™. Greater than 80% of these cells will be cTnT+. An average of 1×10⁶ cells can be harvested from a single well of a 12-well plate. STEMdiff™ cardiomyocyte maintenance kit comprises the maintenance basal medium and supplement; it can be used for long-term maintenance of hPSC-derived cardiomyocytes for one month or longer. These cardiomyocytes can be used in various downstream applications and analyses.
NOTE: This product was formerly named STEMdiff™ cardiomyocyte differentiation kit; the product itself and manufacturing procedures have not changed, but the name has been updated to more accurately reflect the cell type generated.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').