Assistent Instrument Brush with Nylon Bristles
Catalog # HECH91124700017
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Matériau des poils:Nylon
- Ø tête de la brosse:18 mm
- Handle material:Plastique
- Longueur totale:170 mm
- Cdt:10
Specifications
About this item
Instrument cleaning brush for use with laboratory glassware.
- Plastic handle
- Nylon bristles