- Type d'anticorps:Primaire
- Nom de l'antigène:keratin 4, keratin 5, keratin 6A, keratin 6B, keratin 8, keratin 10, keratin 13, keratin 18
- Symbole de l'antigène:KRT4, KRT5, KRT6A, KRT6B, KRT8, KRT10, KRT13, KRT18
- Clonalité:Monoclonal
- Clone:SPM583
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Hôte:Mouse
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Réactivité:Marmoset,Frog,Pig,Goat,Human,Bovine,Guinea pig,Rat,Mouse
- Western blot:Yes
- Format:Purified Antibody
- Formulaire:Liquid
- ID de gène:P19013, P13647, P02538, P04259, P48668, P05787, P13645, P13646, P05783
- Synonymes antigène:KRT4, KRT5, KRT6A, KRT6B, KRT8, KRT10, KRT13, KRT18
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Température de stockage:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Température de transport:4 °C
- Immunogène:A keratin-enriched preparation from cultured human A431 cells was used as the immunogen for this anti-Multi-Cytokeratin antibody.
- Purification:Protein G affinity chromatography
- Taille:100 µg
- Cdt:100 µG
This mAb recognises cytokeratin 4, 5, 6, 8, 10, 13, and 18. This is a broad-spectrum antibody which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the anti-Multi-Cytokeratin antibody for each application should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: KRT4, KRT5, KRT6A, KRT6B, KRT8, KRT10, KRT13, KRT18
Clonality: Monoclonal
Clone: SPM583
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Rat, Mouse, Guinea Pig, Frog, Goat, Marmoset, Pig