Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:CD44 molecule (Indian blood group)
- Symbole de l'antigène:CD44
- Clonalité:Monoclonal
- Clone:SPM544
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Hôte:Mouse
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Réactivité:Baboon,Green monkey,Human
- Western blot:Yes
- Neutralisation:Yes
- Format:Purified Antibody
- Formulaire:Liquid
- ID de gène:P16070
- Synonymes antigène:CD44
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Température de stockage:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Température de transport:4 °C
- Immunogène:Stimulated human leukocytes were used as the immunogen for this anti-CD44 antibody.
- Tested applications:Functional
- Purification:Protein G affinity chromatography
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
Recognises a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes. Its epitope is resistant to digestion by trypsin and chymotrypsin. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85 to 95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion and stromal binding. CD44 immunostaining is commonly used for the discrimination of urothelial transitional cell carcinoma in-situ from non-neoplastic changes in the urothelium.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Functional Studies: Order Ab without BSA/azide Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,25-0,5 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the anti-CD44 antibody for each application should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: SPM544
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Baboon, Green Monkey