Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:SMT3 suppressor of mif two 3 homolog 2 (S. cerevisiae)
- Symbole de l'antigène:SUMO2
- Clonalité:Monoclonal
- Clone:SPM572
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Hôte:Mouse
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Format:Purified Antibody
- Formulaire:Liquid
- ID de gène:P61956
- Synonymes antigène:SUMO2
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Température de stockage:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Température de transport:4 °C
- Immunogène:Recombinant human protein was used as the immunogen for this SUMO-2/3 antibody.
- Purification:Protein G affinity chromatography
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
This mAb reacts with both SUMO-2 and SUMO-3. The small ubiquitin-related modifier (SUMO) proteins, which include SUMO-1, 2 and 3, belong to the ubiquitin-like protein family. Like ubiquitin, the SUMO proteins are synthesised as precursor proteins that undergo processing before conjugation to target proteins. Also, both utilize the E1, E2 and E3 cascade enzymes for conjugation. However, SUMO and ubiquitin differ with respect to targeting. Ubiquitination predominantly targets proteins for degradation, whereas sumoylation targets proteins to a variety of cellular processing, including nuclear transport, transcriptional regulation, apoptosis and protein stability. The unconjugated SUMO-1, 2 and 3 proteins localize to the nuclear membrane, nuclear bodies and cytoplasm, respectively. SUMO-1 utilizes Ubc9 for conjugation to several target proteins, which include MDM2, p53, Pml and RanGap1. SUMO-2 and 3 contribute to a greater percentage of protein modification than does SUMO-1 and unlike SUMO-1, they can form polymeric chains. In addition, SUMO-3 regulates beta-Amyloid generation and may be critical in the onset or progression of Alzheimer s disease.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 0,5 - 1 µg/ml Western blot: 0,5 - 1 µg/ml Immunohistochemistry (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the SUMO-2/3 antibody for each application should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: SUMO2
Clonality: Monoclonal
Clone: SPM572
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human,