Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Multi Cytokeratin 4/5/6/8/10/13/18 Antibody [C11]
- Symbole de l'antigène:CK4, CK5, 6, 8, 10, 13, and 18
- Clonalité:Monoclonal
- Clone:C11
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Hôte:Mouse
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Réactivité:Marmoset,Frog,Pig,Goat,Human,Bovine,Guinea pig,Rat,Mouse
- Western blot:Yes
- Format:Purified Antibody
- Formulaire:Liquid
- ID de gène:3851 (CK4); 3852 (CK5); 3853 (CK6); 3856 (CK8); 3858 (CK10); 3860 (CK13); 3875 (CK18)
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Température de stockage:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Température de transport:4 °C
- Immunogène:A keratin-enriched preparation from cultured human A431 was used as the immunogen for this Multi Cytokeratin antibody.
- Purification:Protein G affinity chromatography
- Taille:100 µg
- Cdt:100 µG
Spécifications
À propos de cet article
This antibody recognises multi cytokeratins: 4, 5, 6, 8, 10, 13, and 18. It is a broad-spectrum cytokeratin antibody which has been reported to be used to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of cytokeratin markers in cancer research and tumor diagnosis.
Flow Cytometry: use the Multi Cytokeratin mAb at 0,5-1 µg/million cells IF: 0,5 - 1 µg/ml WB: 0,5 - 1 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Multi Cytokeratin antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: Multi Cytokeratin 4/5/6/8/10/13/18 Antibody [C11]
Clonality: Monoclonal
Clone: C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Rat, Mouse, Guinea pig, Frog, Goat, Marmoset, Pig