Anti-COX5B Rabbit Polyclonal Antibody
PRSI18-748
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Cytochrome c oxidase subunit 5B, mitochondrial
- Symbole de l'antigène:COX5B
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Précipitation immunologique:Yes
- Isotype:IgG
- Western blot:Yes
- Formulaire:Liquid
- ID de gène:1329
- Synonymes antigène:Cytochrome c oxidase polypeptide Vb|COX5B|mitochondrial|Cytochrome c oxidase subunit 5B
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Température de stockage:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Température de transport:20
- Immunogène:Recombinant fusion protein containing a sequence corresponding to amino acids 32-129 of human COX5B (NP_001853.2).
- Purification:Affinity purification
- Type de conditionnement:Vial
- Cdt:50 µl
Type: Primary
Antigen: COX5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat