Anti-OR4M2 Rabbit Polyclonal Antibody
N° de catalogue PRSI59-712
Fournisseur: ProSci Inc.
New Product
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Olfactory receptor 4M2
- Symbole de l'antigène:OR4M2
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Western blot:Yes
- Épitope:243 - 271 aa
- Formulaire:Liquid
- ID de gène:390538
- Modification:None
- Molecular weight:35 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,45 mg/ml
- Température de transport:4°C
- Immunogène:This OR4M2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 243 - 271 amino acids from the C-terminal region of human OR4M2.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Cdt:400 µl
Spécifications
À propos de cet article
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: OR4M2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 243-271
Host: Rabbit
Isotype: Ig
Reactivity: Human