Anti-ADRB2 Recombinant Antibody [Clone: 1B1.3a]
Référence PRSI24-730
Fournisseur: ProSci Inc.
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:adrenoceptor beta 2
- Symbole de l'antigène:ADRB2
- Clonalité:Recombinant
- Clone:1B1.3a
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Isotype:IgG1 kappa
- Réactivité:Mouse
- Adsorption croisée:No
- Formulaire:Liquid
- ID de gène:16154
- Synonymes antigène:Interleukin-10 receptor subunit|CD210|Interleukin-10 receptor subunit alpha|CDw210a|IBI-3A|IL-10R subunit alpha|IL-10 receptor subunit alpha| Il10ra Il10r|1B1-3A|IL-10R subunit 1|Interleukin-10 receptor subunit 1|IL-10RA|IL-10R1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Température de stockage:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Température de transport:Shipped on blue ice
- Immunogène:This antibody was raised by immunising rats with the purified recombinant extracellular region of mouse Interleukin 10 Receptor
- Tested applications:Blocking
- Type de conditionnement:Vial
- Cdt:0,2 mg
Spécifications
À propos de cet article
Type: Primary
Antigen: adrenoceptor beta 2
Clonality: Recombinant
Clone: 1B1.3a
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Mouse