Hei-TORQUE Overhead Stirrers, Gold 1 Package
N° de catalogue HEID501-61029-04
Fournisseur: Heidolph Instruments GmbH & Co.KG
Spécifications
- Désignation:Hei-TORQUE Gold 1 package, CH-plug
- Seq:03
- Cdt:1
Spécifications
À propos de cet article
In order to provide the perfect end-to-end solution for powerful stirring and the simplest operation in the laboratory, we have added packages to the Hei-TORQUE product group.Each package contains a telescopic stand and a suitable clamp for easy use on lab tables.
Livraison: Hei-TORQUE Ultimate 100 comes with telescope stand and clamp.