Liquid Amies, Sterile, Transporter Swab
Catalog # DELT300284.SE
Supplier: DELTALAB
Specifications
- Stérile:+
- Désignation:Shaft material: snappable PS, tip material: viscose
- Milieu de transport:Liquide Amies
- Capacité de flacon:0,0550 m³
- Cdt:1.200
Specifications
About this item
This is a variation of the traditional Amies media swab where the media is presented on liquid form without agar.
- Peel-pack dimensions: 38×210 mm
Media is supplied in its liquid form, in a sponge, suitable for direct extensions on slides.
Swabs are sterilised by radiation.
Suitable for conservation of pathogens such as:Haemophilus sp., Corynebacterium sp., Streptococcus pyogenes, Streptococcus pneumoniae, Shigella flexneri, Salmonella typhi, Brucella abortus, Staphylococcus epidermidis, Escherichia coli.
Certifications: CE MDD Class IIa. CE IVD.
Caution: Expiry date: 30 months after sterilisation date.