Tétrachlorozincate de 1,3-diamino-7-(diéthylamino)-4-méthylphénoxazine-5-ium (1:2) solution R dans l'éthanol pour la microscopie
Catalog # 16035-100ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Cdt:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: CI 51010, 1,3-Diamino-7-(diethylamino)-3-methylphenoxazin-5-ium tetrachlorozincate (2:1), Brilliant cresyl blue zinc chloride double salt, Brilliant Blue C, BCB
:
:801.96 g/mol
Density:0,808 g/ml at 20 °C
:M-Clarity™ quality level = MQ100
Flash Pt:15 °C (closed cup)
EINECS:200-578-6
:GHS02
:GHS07
Storage Temperature:Room Temperature
CAS Number:81029-05-2
:10127-36-3
Formula:C₃₄H₄₂Cl₄N₈O₂Zn
MDL Number:MFCD00148901