Kidney, nephron and kidney capsule
763-0124
- Désignation:Kidney, nephron and kidney capsule
- Cdt:1
Three models on a common plate. Kidney in cross section enlarged 3×. Nephron enlarged 120× and kidney capsule enlarged 700×. Dimensions: 29×52×9 cm.