ReadiUse™ Spin Filter (5), 10KD
Catalog # AATB60502
Supplier: AAT Bioquest
Specifications
- Désignation:ReadiUse™ 10KD spin filter
- Cdt:5
Specifications
About this item
ReadiUse™ 10 Kd spin filter is a disposable ultrafiltration device for the concentration of biological samples such as antibodies.
- Compatible with both swing bucket or fixed angle instruments
- Designed to prevent membrane fouling
- Low internal surface area
- High material recovery from dilute solutions
Caution: For research use only.