ReadiUse™ Tyramide (TSA)/Styramide (PSA) Optimized Reaction Buffer (1000 Reactions)
Catalog # AATB45090
Supplier: AAT Bioquest
CAS Number:
Specifications
- Cdt:1.000 Tests
- Désignation:ReadiUse™ tyramide (TSA)/styramide (PSA) optimized reaction buffer
Specifications
About this item
For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.
- Improved fluorescent signal
- Suitable for either TSA or PSA reactions
- Ready to use
Caution: For research use only.