ReadiLink™ Rapid iFluor® Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Référence AATB1227
Fournisseur: AAT Bioquest
Numéro CAS:
Spécifications
- Cdt:2 Tests
- Désignation:ReadiLink™ rapid iFluor™ 555 antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'
Spécifications
À propos de cet article
AAT Bioquest's iFluor® dyes are optimised for labelling proteins, in particular, antibodies.
- Red-laser excitable
- Robust and simple antibody labelling protocol
- No column purification required
Attention: For research use only.