Portelite™ Fluorimetric Protein Quantitation Kit, Optimized for CytoCite™ and Qubit™ Fluorometers
Catalog # AATB11111
Supplier: AAT Bioquest
CAS Number:
Specifications
- Cdt:500 Tests
- Désignation:Portelite™ fluorimetric protein quantitation Kit *Optimized for CytoCite™ and Qubit™ Fluorometers*
Specifications
About this item
Protein quantification is an essential task in protein purification, electrophoresis, cell biology, molecular biology and other research applications.
- Dynamic range from 12.5 ug/ml to 5 mg/ml of bsa
- Optimised for cytocite™ and qubit™ fluorometers
- Highly sensitive
- Robust protocol
Caution: For research use only.