ContactUsLinkComponent
ReadiScript™ RT Reverse Transcription Kit
ReadiScript™ RT Reverse Transcription Kit
Catalog # AATB60100
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ReadiScript™ RT Reverse Transcription Kit
Catalog # AATB60100
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Cdt:
    50 Tests
  • Désignation:
    Convenient tool for cDNA synthesis used in gene expression analysis with qPCR
Specifications
About this item
The two-tube kit format helps to set up the reaction faster and is optimised to yield cDNA over a broad dynamic range.
  • Robust protocol
  • Broad dynamic range
  • Useful for gene expression analysis
The 5X supermix contains a unique blend of oligo(dT) and random primer mix to prevent the 5' and 3' bias of the target genes. The reaction mix contains a potent blend of RNase inhibitors to protect RNA during the setup and reverse transcription process..
The resulting cDNA product is compatible with all qPCR chemistries or conventional end-point RT-PCR.
Packaging: Plastic vial.
Caution: For research use only.