Order Entry
Switzerland
ContactUsLinkComponent
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Ofloxacin
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Ofloxacin
Référence AATB50021
Fournisseur:  AAT Bioquest
Numéro CAS:  
undefined
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Ofloxacin
Référence AATB50021
Fournisseur:  AAT Bioquest
Numéro CAS:  

Spécifications

  • Type de conditionnement:
    Vial
  • Cdt:
    1 mg
  • Conjugaison:
    Ofloxacin
  • Type de protéine/peptide:
    Native
  • Source:
    Serum
  • Species:
    Megathura crenulata
  • Synonyme de protéine:
    KLH
  • Nom de la protéine/du peptide:
    Keyhole limpet hemocyanin

Spécifications

À propos de cet article

Ofloxacin-KLH Conjugate is used for developing anti-ofloxacin antibodies that are typically used in an ELISA assay.

  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Attention: For research use only.