Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Fentanyl
Référence AATB50566
Fournisseur: AAT Bioquest
Numéro CAS:
Spécifications
- Type de conditionnement:Vial
- Cdt:5 mg
- Conjugaison:Fentanyl
- Type de protéine/peptide:Native
- Source:Serum
- Species:Megathura crenulata
- Synonyme de protéine:KLH
- Nom de la protéine/du peptide:Keyhole limpet hemocyanin
Spécifications
À propos de cet article
Small compounds (drugs or peptides) are not sufficiently complex by themselves to induce an immune response or be processed in a manner that elicits production of specific antibodies.
- High immunogenicity
- Useful for antibody development
- Fentanyl test building block
Attention: For research use only.