Native Human Concanavalin A (from Human), iFluor® 555
AATB25585
:
- Type de conditionnement:Vial
- Cdt:1 mg
- Conjugaison:iFluor® 555
- Type de protéine/peptide:Native
- Species:Human
- Synonyme de protéine:ConA
- Nom de la protéine/du peptide:Concanavalin A
Concanavalin A (ConA) is a lectin that binds specifically to certain structures found in various sugars, glycoproteins and glycolipids.
- Well-cited conjugate
- High affinity and selectivity
- Multiple applications
: For research use only.