Pluronic® F-127 Cell Culture Tested
N° de catalogue AATB20050
Fournisseur: AAT BIOQUEST
Spécifications
- Cdt:10 g
Spécifications
À propos de cet article
Pluronic® F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility.
- Non-toxic to cells
It is soluble at low temperature, but gels at room temperature.