Cell Meter™ Live Cell Caspase 8 Binding Assay Kit *Red Fluorescence*
Référence AATB20116
Fournisseur: AAT Bioquest
Numéro CAS:
Spécifications
- Cdt:25 Tests
- Désignation:Cell Meter™ Live cell Caspase 8 binding assay kit *Red fluorescence*
- Tests par set:25 Tests
Spécifications
À propos de cet article
Our Cell Meter™ live cell caspases activity assay kits are based on fluorescent FMK inhibitors of caspases.
- Based on FMK inhibitor
- Direct caspase detection
- Suitable for all major fluorescence platforms
Attention: For research use only.