Cell Meter™ Fixable Viability Dye, RX780
Cell Meter™ RX780 fixable cell stain, 200 Tests, 200 Tests
N° de catalogue AATB22536
Fournisseur: AAT Bioquest
Cell Meter™ Fixable Viability Dye, RX780
N° de catalogue AATB22536
Fournisseur: AAT Bioquest
Numéro CAS:
Aller à
Product Details & Documents
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
- Cell-Impermeable
- Amine- And Thiol-Reactive
- High Stability
- Live/Dead Cell Labeling Dye
- Fixable
Caution: For research use only.
Spécifications
Spécifications
- Cdt:200 Tests
- Désignation:Cell Meter™ RX780 fixable cell stain
- Tests par set:200 Tests