ThiolTrace™ Violet, 500
ThiolTrace™ Violet 500, 500 Tests, 500 Tests
N° de catalogue AATB22280
Fournisseur: AAT Bioquest
Aller à
Product Details & Documents
The subcellular detection and localization of GSH is important in understanding the modulation of redox status, the effect of drugs, and the mechanisms of detoxification.
- Extremely Bright Fluorescence
- Compatible With Serum-Containing Media
- High Sensitivity
Attention: For research use only.
Spécifications
Spécifications
- Cdt:500 Tests
- Désignation:ThiolTrace™ Violet 500
- Tests par set:500 Tests
- Cat. No.:AATB22280
- Purity:95%