Wooden Clamp for Test Tubes
NEOLDT-0771
: NEOLAB
- Pour:Test tubes
- Longueur:190 mm
- Matière:Bois
- Cdt:1
Wooden clamp for test tubes.
- Non-sterile
- Non autoclavable
Diameter: 30 mm
Transported in room temperature.
Wooden clamp for test tubes.
Diameter: 30 mm
Transported in room temperature.