Specifications
- Désignation:Sterillium® med hand disinfectant, clear colourless, density (at 20 ℃) approximately 0,82 g/cm³
- Cdt:1 L
Specifications
About this item
Sterillium® med the hand disinfectant with virucidal efficacy within hygienic hand disinfection. Featuring the proven Sterillium® skincare complex. Colourant- and fragrance-free. Flashpoint 20 ℃.
- Comprehensively efficacy, including virucidal activity with application time for hygienic hand disinfection
- Increases skin hydration, preserves skin elasticity and the natural smoothness of the skin
- All-year-product with quick efficacy
- High skin tolerability
- Maintains the natural pH value of the skin
- Colourant- and fragrance-free
- With proven skin care complex
- Composition - Active substances per 100 g: Ethanol 85 g
Sterillium® med is suitable for hygienic and surgical hand disinfection. Sterillium® med is free from colourants and fragrances and thus is particulary suitable for users with sensitive skin. Thanks to its highly efficient formula, Sterillium® med provides full virucidal activity including Norovirus within hygienic hand disinfection and thus is ideally suitable for year round infection prevention.
Microbiology: Bactericidal, fungicidal, tuberculocidal, mycobactericidal, virucidal against enveloped viruses, limited spectrum of virucidal activity including norovirus, virucidal.
Stability: In opened, and tightly closed container, with mounted single-use pump or when using the Eurodispenser 1 plus/1 plus touchless: 12 months.