ContactUsLinkComponent
Gas cartridge, CP250
Gas cartridge, CP250
Catalog # CAMP2000033971
Supplier:  CAMPING GAZ
undefined
Gas cartridge, CP250
Catalog # CAMP2000033971
Supplier:  CAMPING GAZ
Specifications
  • Désignation:
    Gas cartridge, CP 250, 220 g capacity, weight: 315 g, Ø×H: 70×200 mm
  • Cdt:
    1
Specifications
About this item
Campingaz® gas cartridge CP250 with safety valve.
  • Filled with high-performance isobutane gas
  • Equipped with high security self-sealing valve
  • Can be disconnected and reconnected even when the cartridge is not empty