MELAcontrol Pro for Stream Sterilizers
Référence MELAME01075
Fournisseur: MELAG Medizintechnik OHG
Spécifications
- Désignation:MELAcontrol Pro
- Cdt:1
Spécifications
À propos de cet article
MELAcontrol Pro with 40 indicator strips is a durable, usable test body for the power-user.
This is a test body for 'Class B' steam sterilizers, which simulate the complicated requirements for sterilizing hollow instruments. This test body is required for batch monitoring of the processing of 'critical B' instruments.
Certifications: EN 867-5 standard.
Informations de commande: The test strips can be re-ordered in a refill package with 250 indicator strips.