Recombinant Human OSM (from E. coli), His-tag
Catalog # ABNOP8997.20UG
Supplier: Abnova
CAS Number:
Specifications
- Cdt:20 µG
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- ID de gène:P13725
- Nom de la protéine/du peptide:OSM
- Sequence:26a.a.-234a.a.