ContactUsLinkComponent
Recombinant Mouse IL5 (from Sf9 cells), His-tag
Recombinant Mouse IL5 (from Sf9 cells), His-tag
Catalog # ABNOP8281.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse IL5 (from Sf9 cells), His-tag
Catalog # ABNOP8281.10UG
Supplier:  Abnova
CAS Number:  
Specifications
  • Cdt:
    10 µG
  • Type de protéine/peptide:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Mouse
  • Tag sequence:
    His-tag
  • ID de gène:
    P04401
  • Nom de la protéine/du peptide:
    IL5
  • Sequence:
    21a.a.-133a.a.