Order Entry
Switzerland
ContactUsLinkComponent
Recombinant Human GDF10 (from E. coli)
Recombinant Human GDF10 (from E. coli)
Catalog # ABNOP8696.20UG
Supplier:  Abnova
CAS Number:  
Recombinant Human GDF10 (from E. coli)
Catalog # ABNOP8696.20UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Cdt:
    20 µG
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Nom de la protéine/du peptide:
    GDF10

Specifications

About this item