Recombinant Human ANGPT1 (from E. coli)
20 µG
N° de catalogue ABNOP8405.20UG
Fournisseur: Abnova
Recombinant Human ANGPT1 (from E. coli)
N° de catalogue ABNOP8405.20UG
Fournisseur: Abnova
Numéro CAS:
Aller à
Product Details & Documents
Spécifications
Spécifications
- Cdt:20 µG
- ID de gène:Q15389
- Cat. No.:ABNOP8405.20UG
- Sequence:20a.a.-498a.a.
- Species:Human
- Nom de la protéine/du peptide:ANGPT1
- Type de protéine/peptide:Recombinant
- Source:E. coli
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



