Auftragserfassung
Switzerland
ContactUsLinkComponent
Recombinant Human TNFRSF18 (from Baculovirus)
Recombinant Human TNFRSF18 (from Baculovirus)
Catalog # ABNOP8401.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Human TNFRSF18 (from Baculovirus)
Catalog # ABNOP8401.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Cdt:
    10 µG
  • Type de protéine/peptide:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Human
  • ID de gène:
    Q9Y5U5
  • Nom de la protéine/du peptide:
    TNFRSF18
  • Sequence:
    26a.a.-162a.a

Specifications

About this item