Order Entry
Switzerland
ContactUsLinkComponent
 
Recombinant Rat EPO (from HEK293 cells), His-tag
Catalog # ABNOP8577.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Rat EPO (from HEK293 cells), His-tag
Catalog # ABNOP8577.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Cdt:
    10 µG
  • Type de protéine/peptide:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Tag sequence:
    His-tag
  • Nom de la protéine/du peptide:
    EPO

Specifications

About this item