0,00 SFr.
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Protéines et Peptides
Recombinant Human FASLG (from E. coli)
Recombinant Human FASLG (from E. coli)
ABNOP8583.20UG
:
Abnova
:
Recombinant Human FASLG (from E. coli)
ABNOP8583.20UG
:
Abnova
:
Cdt:
20 µG
Type de protéine/peptide:
Recombinant
Source:
E. coli
Species:
Human
Nom de la protéine/du peptide:
FASLG