Recombinant Human FGF14 (from E. coli), His-tag
Catalog # ABNOP8613.10UG
Supplier: Abnova
CAS Number:
Specifications
- Cdt:10 µG
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His-tag
- Nom de la protéine/du peptide:FGF14