0,00 SFr.
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Protéines et Peptides
Recombinant Human IL23A (from E. coli), His-tag
Recombinant Human IL23A (from E. coli), His-tag
ABNOP8111.100UG
:
Abnova
:
Recombinant Human IL23A (from E. coli), His-tag
ABNOP8111.100UG
:
Abnova
:
Cdt:
100 µG
Type de protéine/peptide:
Recombinant
Source:
E. coli
Species:
Human
Tag sequence:
His-tag
Nom de la protéine/du peptide:
IL23A