Recombinant Mouse VCAM1 (from Baculovirus), His-tag
ABNOP7959.250UG
: Abnova
:
- Cdt:250 µG
- Type de protéine/peptide:Recombinant
- Source:Baculovirus
- Species:Mouse
- Tag sequence:His-tag
- ID de gène:Q3UPN1
- Nom de la protéine/du peptide:VCAM1
- Sequence:25a.a.-698a.a.