Order Entry
Switzerland
ContactUsLinkComponent
 
Recombinant Human FGF3 (from E. coli), His-tag
  ABNOP7845.20UG
 :  Abnova
 :  
Recombinant Human FGF3 (from E. coli), His-tag
  ABNOP7845.20UG
 :  Abnova
 :  

 

  • Cdt:
    20 µG
  • Type de protéine/peptide:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Nom de la protéine/du peptide:
    FGF3