Recombinant Human OSM (from E. coli)
Catalog # ABNOP7375.10UG
Supplier: Abnova
CAS Number:
Specifications
- Cdt:10 µG
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Human
- ID de gène:P13725
- Nom de la protéine/du peptide:OSM
- Sequence:26a.a.-234a.a