Recombinant Human HGF (from E. coli)
Catalog # ABNOP7469.5UG
Supplier: Abnova
CAS Number:
Specifications
- Cdt:5 µG
- Type de protéine/peptide:Recombinant
- Source:E. coli
- Species:Human
- ID de gène:P14210
- Nom de la protéine/du peptide:HGF
- Sequence:23a.a.-99a.a.